Would you like to log in again?
Safco® Precision Four-Post Drafting Table Base
Supplier: Safco
Safco®
SAF3962GR
500000-498EA 1107.88 USD InStock
500000-498
Safco® Precision Four-Post Drafting Table Base
Furniture Tables and Workbenches
A professional-quality table for professional-quality results.
  • Stabilizing bar for added support
Industrial-grade steel with a baked enamel finish. Angle of board adjusts up to 50° without tools.
Ordering information: Top and Base sold and shipped separately - order both.
Delivery information: Includes 12¹/₂W × 29³/₄D locking tool drawer and 38¹/₄W × 28³/₄D reference drawer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty