Would you like to log in again?
Quartet® Prestige Plus™ Magnetic Fabric Bulletin Boards, Essendant
Supplier: Essendant
Quartet® Prestige Plus™
QRTMB544M QRTMB544A QRTMB543M QRTMB547A QRTMB543A QRTMB547M
500001-765EA 340.34 USD InStock
500001-765 500001-764 500001-766 500001-783 500001-782 500001-763
Quartet® Prestige Plus™ Magnetic Fabric Bulletin Boards, Essendant
Presentation Boards and Screens
Capture attention and heighten the power of important communications with the Quartet® Prestige Plus® Magnetic Fabric Bulletin Board
Design meets premium function in this sophisticated communication tool ideal for high profile offices and conference rooms. The versatile, magnetic bulletin surface secures documents with pins or magnets. Backed by high-density fiberboard, these top-of-the-line boards hold pins tightly in place so documents stay securely posted. The durable fabric will not crumble or fade like traditional cork boards. Fasten photos or important documents without holes using the included Quartet® Quick Clips and Magnets®. A patented “Easy Mount” hanging system allows for quick and secure installation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty