Would you like to log in again?
Quartet® Total Erase® Magnetic Dry Erase Board, Essendant
Supplier: Essendant
Quartet® Total Erase®
QRT2548 QRT2545 QRT2543 QRT2544 QRT2547
500002-152EA 369.39 USD InStock
500002-152 500002-151 500002-157 500002-156 500002-154
Quartet® Total Erase® Magnetic Dry Erase Board, Essendant
Presentation Boards and Screens
Highly durable and dependable, the Total Erase® board delivers the benefits of a premium porcelain surface
Built to perform in heavy use environments such as busy offices and classrooms. The DuraMax® Porcelain will not scratch or dent. The smooth, easy-to-maintain Total Erase® surface will not stain or ghost, keeping it looking new, even after years of use. The steel-backed porcelain creates a magnetic surface, doubling its use as a bulletin board for additional communication and collaboration options. Includes four Quartet® dry erase markers and mounting hardware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty