Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alera® Valencia Series Bookcase, Essendant LLC MS

Supplier: Essendant
Alera®
ALEVA634432MY ALEVA638232MC ALEVA633032MC ALEVA633032MY ALEVA638232MY ALEVA636632MY ALEVA634432MC ALEVA636632MC ALEVA635632MC ALEVA635632MY
500002-659EA 279.43 USD InStock
500002-659 500002-669 500002-658 500002-668 500002-695 500002-661 500002-694 500002-660 500002-799 500002-798
Alera® Valencia Series Bookcase, Essendant LLC MS
Book Holders
Woodgrain laminate bookcase features a contemporary design that fits perfectly in professional environments.

  • Dent-, scratch- and water resistant
  • Tight, dowel and cam-lock assembly provides a clean look while ensuring overall stability
  • 3 mm PVC protective edge banding
  • 11.75" deep adjustable shelves accommodate a wide range of materials
  • Top panel is 14.5" deep for door installation
  • Shelf Capcity: 150 lbs.

Shelf thickness 1".

Certifications: GREENGUARD Indoor Air Quality Certified®, Meets or exceeds ANSI/BIFMA standards.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty