Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Victor® 1190 Executive Desktop Calculator
Supplier: Victor
Victor®
VCT1190
500003-747EA 31.2 USD InStock
500003-747
Victor® 1190 Executive Desktop Calculator
Calculators Desktop Calculators
PC touch keyboard with large contoured keys for comfort and more accurate entries.
  • Wall mountable
  • Tilted LCD displays 12 digits (16 mm height) with punctuation for easy reading
  • Dual solar/battery power for use in any lighting condtion
  • Auto power off
Features decimal function, tax calculation, percent key, change sign (+/–) key, backspace key, double zero key, cost/sell/margin calculation, square root key, percent add-on/discount and 3-key memory.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty