Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Canon® Toner Cartridge, 2659B001, 2660B001, 2661B001, 2662B001, Essendant LLC MS

Supplier: Essendant
Canon®
CNM2662B001 CNM2661B001 CNM2659B001 CNM2660B001
500004-617EA 212.71 USD InStock
500004-617 500004-616 500004-615 500004-743
Canon® Toner Cartridge, 2659B001, 2660B001, 2661B001, 2662B001, Essendant LLC MS
Inks
Cartridges are dependable and long-lasting.

  • 5% coverage
  • OEM and no remanufacturing available

Brings out the best in your text and images. Easy to install and use.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty