Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HON® 2090 Pillow-Soft® Series Executive High-Back Swivel/Tilt Leather Chair

Supplier: HON
HON2091SR11T HON2091SR69T
500005-162EA 714.45 USD InStock
500005-162 500005-161
HON® 2090 Pillow-Soft® Series Executive High-Back Swivel/Tilt Leather Chair
Chairs
The executive swivel chair features a memory foam seat cushion which responds to body dynamics and a fiber fill in back with recess at the base to cradle the lumbar region.

  • Tufted, gathered cushions with stain-resistant protection
  • Loop style arm rests
  • 360° swivel, tilt with tilt lock and tension
  • Pneumatic seat height adjustment
  • Distance between arms 20.5"
  • Hard wheel tread

Five star base with five casters.

Certifications: Meets or exceeds ANSI/BIFMA standards, Scientific Certification Systems (SCS) Certified Indoor Advantage™ Gold

Ordering information: Casters supplied with this chair are not suitable for all floor types.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty