Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alera® Epoch Series Mesh Mid-Back Swivel/Tilt Multifunction Chair

Supplier: Alera
Epoch
ALEEP42ME10B
500005-507EA 458.95 USD InStock
500005-507
Alera® Epoch Series Mesh Mid-Back Swivel/Tilt Multifunction Chair
Chairs
Aesthetically designed 360° Swivel multifunctional chair for contemporary appeal.

  • Mesh-covered curved back for added support
  • Integral lumbar support
  • Pneumatic seat height adjustment
  • Height-adjustable backrest, tiltable with tilt lock and tension
  • Variable synchro-tilt
  • Height- and width-adjustable arms with black soft polyurethane pads
  • Hard wheel tread

Five-star base with 5 casters.
Arm height from seat 6.75 - 9.25"
Distance between arms 21.25"

Certifications: GREENGUARD Indoor Air Quality Certified®, Meets or exceeds ANSI/BIFMA standards

Ordering information: Casters supplied with this chair are not suitable for all floor types.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty