Would you like to log in again?
Bankers Box® R-KIVE® Maximum Strength Storage Boxes
Supplier: Bankers Box
Bankers Box® R-KIVE®
FEL12775 FEL0724303 FEL00724 FEL00725 FEL0083601 FEL0072506
500010-742CT 199.56 USD InStock
500010-742 500010-741 500010-015 500010-685 500010-746 500010-745
Bankers Box® R-KIVE® Maximum Strength Storage Boxes
Boxes Storage Boxes
Heavy duty, triple-end, double-side construction stacks up stronger and withstands frequent handling
FastFold™ quick and easy assembly. Reinforced tear-resistant hand holes make box comfortable to carry. Deep, locking lift-off lid stays in place for secure file storage. Smooth rolled edges add strength and prevent paper cuts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty