Would you like to log in again?
Memphis™ Dyneema® Gloves, Essendant
Supplier: Essendant
Memphis™ Dyneema®
CRW9672XL CRW9672L
500012-832PR 13.08 USD InStock
500012-832 500012-835
Memphis™ Dyneema® Gloves, Essendant
Gloves
The economical choice for solid protection against cuts and abrasions.
  • Flexible material remains breathable and lint free
  • Laundry safe and easy to clean, with no loss in cut resistance
Polyurethane palm and finger coating provides a sturdy, durable grip in both wet and dry applications.
Certifications: EN 388 (4341)
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty