Would you like to log in again?
Memphis™ Ninja® HPT Gloves, Essendant
Supplier: Essendant
Memphis™ Ninja®
CRWN9699L CRWN9699S CRWN9699XL CRWN9699M
500012-932PR 3.58 USD InStock
500012-932 500012-922 500012-926 500012-938
Memphis™ Ninja® HPT Gloves, Essendant
Gloves
When working in both wet and dry environments, professional-grade gloves maintain a solid, durable performance.
  • Gloves are made of nylon and silicone
  • CE Score: 4131
Hydropellent technology coating process creates encapsulated air molecules which repel liquids for a firm wet or dry grip. Thick coating provides a cushioning effect to minimize impact. Gloves remain fresh and free of bacteria with Actifresh® coating. Great for assembly, plumbing, shipping, and general maintenance applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty