Would you like to log in again?
Memphis™ Flex Latex Gloves, Essendant
Supplier: Essendant
Memphis™
CRW96731S CRW96731M CRW96731L CRW96731XL
500012-923PR 2.73 USD InStock
500012-923 500012-945 500012-935 500012-929
Memphis™ Flex Latex Gloves, Essendant
Gloves
Multi-purpose work gloves provide durable strength and grip, without compromising breathability.
  • CE Score: 3131
Gray nylon shell offers added comfort and flexibility. Blue foam latex-dipped palm and fingertips maintain a solid grip in both wet and dry environments. Ideal for small parts handling where tactile sensitivity and dexterity are important.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty