Would you like to log in again?
Stanley Bostitch® GlueShot™ Dual Melt™ Glue Gun
Supplier: Stanley Bostitch
Stanley Bostitch® Glue Shot™ Dual Melt™
BOSGR252
500012-961EA 22.38 USD InStock
500012-961
Stanley Bostitch® GlueShot™ Dual Melt™ Glue Gun
Adhesives
The Stanley Bostitch® GlueShot™ Dual Melt™ Glue Gun offers the versatility of both high and low temperature gluing in one gun.
  • Glue Gun uses hot melt, standard dual-temperature glue
  • Convenient high/low selector switch
  • Trigger feed and nozzle check valve for even application
  • Heats up quickly and bonds within 60 seconds
  • Fold-out stand for safe, handy storage while in use
High temperature is perfect for bonding ceramic, plastic, and metal. Low temperature is better suited for delicate or flammable material such as paper, lace, foil, foam, and lightweight fabric. Uses standard dual-temperature glue sticks on either setting or hot melt on the high setting. For use with Glue Stick BOSGS15, Glue Stick BOSGS20, Glue Stick BOSGS230, Glue Stick BOSGS25, Glue Stick BOSGS260, or Glue Stick BOSGS500.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty