Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Canon® Ink Tank, BCI6BK, BCI6C, BCI6M, BCI6PC, BCI6PM, BCI6Y, Essendant LLC MS

Supplier: Essendant
Canon®
CNMBCI6C CNMBCI6PM CNMBCI6M CNMBCI6PC CNMBCI6Y
500014-359EA 13.43 USD InStock
500014-359 500014-355 500014-356 500014-357 500014-358
Canon® Ink Tank, BCI6BK, BCI6C, BCI6M, BCI6PC, BCI6PM, BCI6Y, Essendant LLC MS
Inks
Each cartridge creates stunning visuals.

  • Cartridge with 5% coverage
  • OEM and not remanufactured

Handles even the most complex tasks. Uses ink reliably and effectively to maximize printing potential.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty