Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xerox® Solid Stick, 108R00668, 108R00669, 108R00670, 108R00671, 108R00672, Essendant LLC MS

Supplier: Essendant
Xerox®
XER108R00670 XER108R00669 XER108R00671
500015-011BX 238.62 USD InStock
500015-011 500015-012 500015-013
Xerox® Solid Stick, 108R00668, 108R00669, 108R00670, 108R00671, 108R00672, Essendant LLC MS
Inks
Brings life to any presentation with striking solids and smooth shades.

  • Clean and easy to use
  • Interlocking ink shapes
  • Vibrant prints
  • OEM and no remanufacturing available
  • Ink stick with 5% coverage

Features unique numbering system for easy loading. Produces vibrant prints on a wide range of media. Tektronix ColorStix® solid inks provide a very appealing cost-per-print, without the expense of dye sublimation output. Just drop them into the shape- and color-coded delivery slots and your printer is back on the job.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty