Would you like to log in again?
BIC® Round Stic® Ballpoint Pen
Supplier: Essendant
BIC® Round Stic®
BICGSM11RD BICGSM11BE BICGSM609BK BICGSM11BK BICGSF11BE BICGSF11BK BICGSM609BE
500020-870DZ 1.92 USD InStock
500020-870 500020-871 500020-864 500020-865 500020-867 500020-868 500020-869
BIC® Round Stic® Ballpoint Pen
Pens Ink Pens
The economical choice for performance and reliability.
  • Lightweight, flexible round barrel for writing comfort
  • Long-lasting: more than 7200 feet of writing in each pen
  • Smooth grip and conical tip
  • With pocket clip
  • Colored frosted cap and translucent barrel for visible ink supply
Not refillable.
Certifications: Ventilated cap meets ISO safety standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty