Would you like to log in again?
Salter Brecknell 100 and 250 lbs. Portable Bench Scales
Supplier: Essendant
SBWGP100 SBWGP250
500024-374EA 125.54 USD InStock
500024-374 500024-366
Salter Brecknell 100 and 250 lbs. Portable Bench Scales
Balances and Scales
Portable for easy transport; features attached handle
  • Tare feature
  • Weight units in pounds
  • Electronic postal rates
Internal, backlit display with one-inch high number readout. Holds weight reading when display is covered by oversize package. Discreet keyboard technology ensures longevity and reliability. Electro-mechanic scale maintains accuracy, even when subjected to high weights. 22-gauge steel base protects internal mechanism from damage. Auto-zero tracking or manual push button returns scale to zero when displayed weight is less than 2% of capacity. Withstands 400-lbs. of weight without damaging scale. Programmable battery life shut-down feature. Black color.
Ordering information: Supplied with 9V battery, an AC adapter is available which must be ordered separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty