Would you like to log in again?
3M™ High-Capacity Maintenance Folded Sorbent
Supplier: 3M
MMMMFL550DD
500025-051BX 68.18 USD InStock
500025-051
3M™ High-Capacity Maintenance Folded Sorbent
Spill Control Materials
The versatile solution for cleaning up spills, drips, and fluid leaks can be used as a boom, pillow, pad, or roll.
  • Increased fluid-holding capacity means less sorbent used; minimizes sorbent waste for disposal
  • Permits simple reclamation for recycling of valuable liquids and reuse of sorbent
  • High-capacity sorbent has a scrim on one side to increase strength and help eliminate linting
  • Perforations every 16" to select the required length
Easy-to-use: Simply pull out and tear off the desired length to use as a boom, unfold once to use as a pillow, or unfold it all the way to use as a pad or roll.
Caution: 3M products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations that accompany each item.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty