Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cosco® Max™ Work Platform

Supplier: Cosco
CellBind®
CSC11880PBLW1
500025-729EA 168.62 USD InStock
500025-729
Cosco® Max™ Work Platform
Ladders and Step Stools
Lightweight yet durable steel step stool ladder has three heavy-duty polypropylene resin steps with slip-resistant treads and a large 14.75x12" top platform step

  • With non-marring, skid-resistant feet to provide stability and protect floor surfaces and easy one-hand fold and lock release
  • Extended hand-grip handle with tool holder

Spread size: 31.5"
Folded depth: 3"
Step WxD: 14.75x12"

Certifications: ANSI Type II, OSHA Compliant

Caution: Metal ladders conduct electricity. Do not use where contact may be made with live electrical circuits.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty