Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iceberg IndestrucTables Too™ 1200 Series Picnic Bench Table

Supplier: Iceberg
IndestrucTables Too™ Iceberg
ICE65923
500026-582EA 643.27 USD InStock
500026-582
Iceberg IndestrucTables Too™ 1200 Series Picnic Bench Table
Furniture Tables and Workbenches
Commercial grade, lightweight, durable and portable table, seats eight, for indoor and outdoor use. Contemporary design, heavy-duty top with radius edges.

  • Blow-molded, high-density polyethylene top is dent-, scratch-, stain-resistant and easily washable
  • Will not fade or crack in sunlight
  • Heavy-gauge, sturdy 1" steel tube legs with charcoal gray powder coated finish
  • 1.5" diameter heavy steel tube base with 0.75" cross bracing
  • Non-marking feet

Capacity load: 600 lbs.; 30×72; Height: 29"

Perfect for indoor/outdoor break areas or parks, this sturdy, weather-resistant table will last for years.

Certifications: Meets or exceeds ANSI/BIFMA standards.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty