Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barricks Utility Table

Supplier: Barricks
BRKUT1860WA BRKUTS1848LQ BRKUT244830WA BRKUTS1848WA BRKUT183030LQ BRKUT183030WA
500026-659EA 272.5 USD InStock
500026-659 500026-645 500026-644 500026-646 500026-643 500026-660
Barricks Utility Table
Furniture Tables and Workbenches
An easy way to expand the working area.

  • Economically priced table with 0.75" woodgrain laminate top
  • Stable nonfolding 1" square steel legs with plastic glides
  • Enamel finish on 2.25" apron and legs

Height: 30"

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty