Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Innovera® MICR Toners, 83027TMICR, 83038TMICR, 83039TMICR, 83061TMICR, 83082TMICR, Essendant LLC MS

Supplier: Essendant
Innovera®
IVR83039TMICR IVR83061TMICR IVR83082TMICR IVR83038TMICR IVR83027TMICR
500027-447EA 234.11 USD InStock
500027-447 500027-444 500027-443 500027-442 500027-441
Innovera® MICR Toners, 83027TMICR, 83038TMICR, 83039TMICR, 83061TMICR, 83082TMICR, Essendant LLC MS
Inks
Innovera® remanufactured toners will save you money compared to nationally branded cartridges. It delivers high-quality performance.

  • Fully backed with a 100% performance guarantee
  • Pay less than name brand printer replacements
  • OEM and remanufacturing available
  • Cartridge with 6% coverage

Engineered to provide optimal efficiency at an economical price. The special high adhesion toner formula, designed specifically for your printer, significantly reduces the possibility of costly fees stemming from rejected checks.

Certifications: Meets ANSI/ABA and all US and Canadian banking standards.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty