Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lexmark™ Toner Cartridge, C544X1YG, C544X1MG, C544X1CG, C544X1KG, Essendant LLC MS

Supplier: Essendant
Lexmark™
LEXC544X1CG LEXC544X1YG LEXC544X1KG LEXC544X1MG
500027-778EA 229.8 USD InStock
500027-778 500027-777 500027-776 500027-779
Lexmark™ Toner Cartridge, C544X1YG, C544X1MG, C544X1CG, C544X1KG, Essendant LLC MS
Inks
These high-capacity, long-lasting cartridges offer a low cost-per-page, with fewer replacements and less downtime.

  • Suitable for laser printers
  • OEM and not remanufactured
  • With return program
  • Extra high-yield
  • Prints at 5% of page coverage

Features a clean installation, so the toner stays in the cartridge and not on your hands. Prints high-quality text, images, and graphics that live up to the source material.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty