Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ricoh® Toner Cartridge, 406347, 406346, 406345, 406344, 406478, 406477, 406476, 406475, Essendant LLC MS

Supplier: Essendant
Ricoh®
RIC406476 RIC406478 RIC406477 RIC406344 RIC406345 RIC406347 RIC406346 RIC406475
500027-789EA 79.26 USD InStock
500027-789 500027-781 500027-792 500027-791 500027-790 500027-783 500027-782 500027-793
Ricoh® Toner Cartridge, 406347, 406346, 406345, 406344, 406478, 406477, 406476, 406475, Essendant LLC MS
Inks
Make the most out of your printing budget with high-quality toner.

  • 5% coverage
  • OEM and no remanufacturing available

Delivers superior performance, with crisp, dark output from first to last page. Easy to install and replace.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty