Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xerox® Toner Cartridges, 113R00689, 113R00690, 113R00691, 113R00692, 113R00693, 113R00694, 113R00695, Essendant LLC MS

Supplier: Essendant
Xerox®
XER113R00691 XER113R00692 XER113R00694 XER113R00690 XER113R00693 XER113R00689
500028-512EA 489.1 USD InStock
500028-512 500028-193 500028-195 500028-164 500028-166 500028-165
Xerox® Toner Cartridges, 113R00689, 113R00690, 113R00691, 113R00692, 113R00693, 113R00694, 113R00695, Essendant LLC MS
Inks
These toner cartridges are utilizing a revolutionary new toner manufacturing process where toner is chemically grown and processed into very small and consistent particles, resulting in sharper, high-gloss image quality.

  • OEM and no remanufacturing available
  • Create striking documents and presentations with ease
  • Print at 5% page coverage
  • Cartridge type 6120 HC are providing high yield than the normal type

Experience enhanced fine-line detail plus superior reliability. Installation is simple and time and cost effective.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty