Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lexmark™ Toner Cartridge, E360H21A, E360H11A, E260A21A, E260A11A, Essendant LLC MS

Supplier: Essendant
Lexmark™
LEXE260A11A LEXE360H11A LEXE260A21A LEXE360H21A
500028-346EA 298.74 USD InStock
500028-346 500028-345 500028-347 500028-344
Lexmark™ Toner Cartridge, E360H21A, E360H11A, E260A21A, E260A11A, Essendant LLC MS
Inks
Creates sharp lines, crisp edges, and smooth shades.

  • Suitable for laser printers
  • OEM and not remanufactured
  • With return program
  • Prints at 5% of page coverage

Highlights the finer details in your work. Each cartridge has been thoroughly tested and inspected to ensure that it is of the highest quality.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty