Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xerox® Toner Cartridge, 106R01433, 106R01434, 106R01435, 106R01436, 106R01437, 106R01438, 106R01439, Essendant LLC MS

Supplier: Essendant
Xerox®
XER106R01437 XER106R01433 XER106R01438 XER106R01436 XER106R01439
500028-791EA 959.63 USD InStock
500028-791 500028-790 500028-793 500028-792 500029-621
Xerox® Toner Cartridge, 106R01433, 106R01434, 106R01435, 106R01436, 106R01437, 106R01438, 106R01439, Essendant LLC MS
Inks
Genuine OEM quality.

  • 5% coverage
  • OEM and no remanufacturing available

Dependable and reliable. Clean and simple installation.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty