Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Innovera® Laser Cartridge, 7551A, 7551MICR, 7551X, Essendant LLC MS

Supplier: Essendant
Innovera®
IVR7551A IVR7551X
500028-966EA 65.78 USD InStock
500028-966 500028-967
Innovera® Laser Cartridge, 7551A, 7551MICR, 7551X, Essendant LLC MS
Inks
Create vivid, vibrant images easily.

  • Cartridge with 5% coverage
  • Compatible and remanufactured

Achieve attention-getting, professional-looking documents. Easy to install.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty