Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lexmark™ Toner Cartridge, X463X11G, X463X21G, X463H21G, X463H11G, X463A11G, Essendant LLC MS

Supplier: Essendant
Lexmark™
LEXX463X11G LEXX463H11G LEXX463A11G
500029-507EA 423.26 USD InStock
500029-507 500029-504 500029-503
Lexmark™ Toner Cartridge, X463X11G, X463X21G, X463H21G, X463H11G, X463A11G, Essendant LLC MS
Inks
Reliability increases productivity and lowers your overall costs of printing.

  • 5% Coverage
  • Return Program
  • OEM and not remanufactured

Creates crisp blacks and sharp edges in every printout. Lexmark™ cartridges provide low-maintenance printing with less downtime.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty