Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xerox® Imaging Units, 108R00647, 108R00648, 108R00649, 108R00650, Essendant LLC MS

Supplier: Essendant

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Xerox®
XER108R00649 XER108R00650 XER108R00647 XER108R00648
500030-288EA 483.15 USD InStock
500030-288 500030-287 500030-289 500030-286
Xerox® Imaging Units, 108R00647, 108R00648, 108R00649, 108R00650, Essendant LLC MS
Inks
This imaging unit is exclusively OEM engineered and manufactured.

  • OEM and no remanufacturing available
  • Prints with 5% coverage

It provides the consistent print quality and performance you demand. Built to stringent specifications for reliable results.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty