Would you like to log in again?
Electrolux Sanitaire® Quick Kleen® Fan Chamber with Vibra Groomer I®
Supplier: Electrolux Sanitaire
Electrolux Sanitaire® Quick Kleen®
EURSC888N
500030-334EA 367.94 USD InStock
500030-334
Electrolux Sanitaire® Quick Kleen® Fan Chamber with Vibra Groomer I®
Vacuum Cleaners
This high-quality commercial vacuum cleaner.
  • Micron Filter® system retains up to 99% of pollens and dust
  • Designed with the Quick Kleen® removable, clear fan chamber that allows for easy access when unit needs service
  • The Vibra Groomer II® chrome steel brush roll is capable of handling the toughest jobs.
  • Six carpet height settings allow you to choose the correct height for a variety of carpets, ideal for large commercial buildings
  • Vinyl bumper, large easy-roll wheels and chrome steel motor hood
  • Utilizes disposable F&G dust bags
Ideal for hospitality industry where high standards of cleanliness need to be maintained at all times.
Replacement bag: EUK-63213A-10
Replacement belt: EUK-52100C-12
Certifications: CRI Green Label, CSA, OSHA approved for commercial use, UL approved
Ordering information: On-Board Tools, Headlight, Furniture Guard, and Dust/Dirt Cup not available.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty