Would you like to log in again?
Safco® Adjustable Height Mini-Tower Computer Workstation, Essendant LLC MS
Supplier: Essendant
Safco®
SAF1927CY
500030-707EA 974.66 USD InStock
500030-707
Safco® Adjustable Height Mini-Tower Computer Workstation, Essendant LLC MS
Furniture Tables and Workbenches
Features 'step-less infinite adjustability' for increased personal comfort and productivity
  • Black powder coated steel frame
  • Adjustable height
  • Adjustable monitor platform height
  • Adjustable printer shelf height in 1" increments
  • 2 locking casters, dual-wheel casters
  • Cable management
  • Safety cap amd safety edge
  • Shelves for 1 CPU, 1 keyboard, 1 monitor and 1 printer
Safety cap with cable management side covers. Includes tool for quick and easy assembly. Monitor and keyboard shelves adjust in 5" range, 29" to 34" high. Printer shelf adjusts in 1" increments.
Keyboard drawer work height - 29 to 33"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty