Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Energizer® e²® NiMH Rechargeable Batteries, Essendant
Supplier: Essendant
Energizer® e²®
EVENH15BP4 EVENH22NBP EVENH12BP4 EVENH35BP2 EVENH50BP2
500031-894PK 25.06 USD InStock
500031-894 500031-892 500031-898 500031-896 500031-900
Energizer® e²® NiMH Rechargeable Batteries, Essendant
Batteries
Rechargeable nickel-metal hydride batteries are engineered to work with your most advanced, high-drain devices such as digital cameras, PDAs, hand-held games, portable CD players and MP3 players.
With a total recharge capacity of 250 instances, these cells truly provide long-lasting power. Cadmium-free construction helps you minimize your environmental footprint.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty