Would you like to log in again?
Fresh Products Conqueror 103 Odor Counteractant Concentrate
Supplier: Essendant
Conqueror 103
FRS1232WBLECT FRS1WBCHCT
500032-418CT 82.09 USD InStock
500032-418 500032-702
Fresh Products Conqueror 103 Odor Counteractant Concentrate
Cleaning Agents Detergents and Surface Cleansers
This water-soluble alkaline formula cleans, deodorizes and refreshes surfaces.
  • General purpose cleaner, applicable on ceramic, fabric, plastic, porcelain, stainless steel and wood
  • Add 1 – 2 oz. per gallon of water or cleaning solution
The formula contains powerful detergents and potent perfumes to counteract malodors and leave the area smelling fresh and clean. Use in fabric shampoo, mop water, any water-based solution. Can be used in trigger sprayers, carpet extractors and laundry.
Certifications: VOC Compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty