Would you like to log in again?
simple green® All-Purpose Industrial Cleaner/Degreaser
Supplier: Essendant
Simple Green®
SMP13005EA
500032-880EA 12.26 USD InStock
500032-880
simple green® All-Purpose Industrial Cleaner/Degreaser
Cleaning Agents Detergents and Surface Cleansers
Strong yet safe cleaner removes oil, lubricants and grime from washable surfaces.
  • Concentrated liquid, dilutes with water and rinses easily
  • Applicable on fiberglass, plastic, stainless steel and wood
  • Biodegradable, nontoxic, nonabrasive and nonflammable
  • Safe for food-preparation areas
Used for pressure washing and steam cleaning, maintenance. Exempt from OSHA and EPA special handling and personal protection requirements for hazardous materials.
Certifications: USDA A4, A8 compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty