Would you like to log in again?
Big D Industries Water-Soluble Deodorant
Supplier: Big D Industries
BGD1618
500033-104CT 95.68 USD InStock
500033-104
Big D Industries Water-Soluble Deodorant
Deodorants and Air Fresheners
Add a small amount of this concentrated liquid formula to water or other cleaning products to destroy nasty odors, without having to worry about altering the cleaner property.
  • Formula is nontoxic, nonflammable, biodegradable and will not stain any material unaffected by plain water
Ideal for mopping, shampooing, rinsing or use in hand sprayer or fogger. Leaves a smooth, lemon fragrance wherever it is used.
Certifications: NSF/USDA D2 compliant
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty