Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sharpie® Industrial Permanent Marker
Supplier: Essendant
Sharpie®
SAN13601
500034-938DZ 36.4 USD InStock
500034-938
Sharpie® Industrial Permanent Marker
Pens Permanent Markers
For industrial, laboratory and commercial use.
  • With fine tip and non-washable ink
Tough under extreme conditions-even at 500 °C. Great for metal, foil, glass, film and plastic. Fade and water-resistant black ink.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty