Would you like to log in again?
Carter's® Felt Stamp Pad, Essendant
Supplier: Essendant
Carter's®
AVE21071 AVE21021 AVE21081 AVE21082
500035-112EA 3.31 USD InStock
500035-112 500035-110 500035-114 500035-108
Carter's® Felt Stamp Pad, Essendant
Stamps and Ink Pads
Felt stamp pad delivers clear, quality impressions.
  • Provides the cushion of foam with the long lasting quality of felt
  • Requires less reinking than foam, holds twice the ink
  • Approximately 15000 impressions before reinking
Reinkable with Neat-Flo inkers to extend the stamp pad's life through thousands more impressions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty