Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Innovera® Laser Cartridge, 83011A, 83011X, Essendant LLC MS

Supplier: Essendant

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Innovera®
IVR83011X IVR83011A
500037-454EA 84.68 USD InStock
500037-454 500028-838
Innovera® Laser Cartridge, 83011A, 83011X, Essendant LLC MS
Inks
Provides reliable toner quality and performance.

  • Cartridge with 5% coverage
  • Compatible and remanufactured

Produces striking black and grayscale printing with optimum results and superior yields. Simple to install and replace.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty