Would you like to log in again?
3M™ Red Buffer Floor Pads 5100
Supplier: 3M
MMM08394
500039-344CT 43.56 USD InStock
500039-344
3M™ Red Buffer Floor Pads 5100
Wipes Cleaning Kits and Pads
Specially designed for spray buffing.
Cleans when damp; buffs when dry. Quickly cleans and removes scuff marks and enhances floor appearance. Works great on automatic floor scrubbers for light-duty cleaning.
Used with rotary or automatic machine speed of 175 to 600 rpm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty