Would you like to log in again?
3M Blue Cleaner Pads 5300
Supplier: 3M
MMM08412 MMM08405
500039-349CT 37.74 USD InStock
500039-349 500039-346
3M Blue Cleaner Pads 5300
Wipes Cleaning Kits and Pads
For heavy-duty scrubbing prior to recoating and spray cleaning.
Removes dirt, spills and scuffs, leaving a clean surface ready for recoating.
Used with rotary or automatic machine speed of 175 to 600 rpm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty