Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Canon® Toner Cartridge, 2789B003AA, 2793B003AA, 2797B003AA, 2801B003AA, Essendant LLC MS

Supplier: Essendant
Canon®
CNM2797B003AA CNM2793B003AA CNM2801B003AA CNM2789B003AA
500040-671EA 168.59 USD InStock
500040-671 500040-672 500040-670 500040-673
Canon® Toner Cartridge, 2789B003AA, 2793B003AA, 2797B003AA, 2801B003AA, Essendant LLC MS
Inks
These OEM quality toner is dependable and reliable.

  • Clean and simple installation
  • Cartridge with 5% coverage

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty