Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AT&T® SynJ™ Expansion Handset, Essendant

Supplier: Essendant
AT&T® Syn J™
ATTSB67108
500041-557EA 188.82 USD InStock
500041-557
AT&T® SynJ™ Expansion Handset, Essendant
Telephones and Headsets
Expand your SynJ™ phone system by adding another handset.

  • Cordless handsets feature access to push-to-talk intercom, paging, call transfer and three-way conferencing
  • 4 line LCD display
  • Built-in speakerphone allows handsfree use
  • Compatible with most headsets (via 2.5 mm jack) and hearing aids

Caller ID and Call Waiting require subscribing to local phone company services. For use with SynJ™ System.

Frequency: 1.9 GHz
Memory/Redial: 10
One Touch Presets: 10
Speed Dial Memories: 100

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty