Would you like to log in again?
Brother® P-Touch® TZ/TZe Series Standard Adhesive Laminated Labeling Tape, Essendant
Supplier: Essendant
Brother® P-Touch®
BRTTZE631 BRTTZE451 BRTTZE355 BRTTZE335 BRTTZE211 BRTTZE641 BRTTZE243 BRTTZE231 BRTTZE232 BRTTZE541 BRTTZE151 BRTTZE334 BRTTZE251 BRTTZE325 BRTTZE241 BRTTZE2312PK BRTTZE242 BRTTZE135 BRTTZE131 BRTTZE141 BRTTZE221 BRTTZE111 BRTTZE345 BRTTZE315 BRTTZE741 BRTTZE121 BRTTZE344 BRTTZE252 BRTTZE145 BRTTZE354 BRTTZE1312PK BRTTZE651 BRTTZE441
500041-785EA 11.83 USD InStock
500041-785 500041-797 500041-798 500041-810 500041-799 500041-812 500041-813 500041-814 500041-815 500041-816 500041-817 500041-818 500041-790 500041-791 500041-792 500041-793 500041-794 500041-795 500041-796 500041-786 500041-787 500041-788 500041-800 500041-789 500041-801 500041-802 500041-803 500041-804 500041-805 500041-806 500041-807 500041-808 500041-809
Brother® P-Touch® TZ/TZe Series Standard Adhesive Laminated Labeling Tape, Essendant
Labels
Perfect for labeling file folders and binders, name badges, and small signage
Laminated surface wipes clean. Resists water, grease, grime, fading and abrasion. Withstands temperatures from -30°C to 100°C, allowing use for outdoor applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty