Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Swingline® Cross-Cut Shredder, EX12-05, Essendant LLC MS

Supplier: Essendant
Swingline®
SWI1757390
500044-516EA 448.62 USD InStock
500044-516
Swingline® Cross-Cut Shredder, EX12-05, Essendant LLC MS
Shredders
The compact sized cross-cut shredder fits nicely under your desk. It is ideal for home office use and makes it easy to protect your personal information.

  • Contain auto start or stop with bin full indicator
  • Shreds DVDs, CDs, credit cards, paper clips, staples
  • Separate feed slot allows for safe shredding of CDs
  • Runs consecutively for 20 minutes with 15 minutes off to ensure longevity of the motor
  • Quiet operation reduces noise disruptions
  • Pull-out bin is easy to empty

Shred size: ⁵⁄₃₂×½"
Sheet capacity: 7
Shredder speed: 8 ft./min
Bin size: 5 gal.
Throat size: 9"

Certifications: (DIN 66399) P-4

Ordering information: Shredder includes wastebin.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty