Would you like to log in again?
Mortars and Pestles, Sapphire, Diamonite™
Supplier: CERCO LLC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
E-14376 E-14377
50405-026EA 1655.75 USD InStock
50405-026 50405-048
Mortars and Pestles, Sapphire, Diamonite™
Mortars and Pestles Mortars with Pestles
Grinds materials not readily reduced in ceramic or metal mortars
Made of a synthetic sapphire substance composed of fine-grained crystals of high-purity aluminum oxide held together by a high strength, non-metallic, vitreous bond. Smooth, but not glossy, hard, pitless grinding surface. Inert to most corrosive chemicals and impervious to liquids under vacuum. Specific gravity, 3.6. Hardness, approximately 9 on Mohs scale.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty