Would you like to log in again?
RheFlex® PEEK™ Fittings, Rheodyne®, IDEX Health & Science
Supplier: IDEX Health & Science
Rheodyne® RheFlex®
6000-051 6000-054 6000-055
50806-932PK 83 USD InStock
50806-932 50806-930 50807-520
RheFlex® PEEK™ Fittings, Rheodyne®, IDEX Health & Science
Fittings and Connectors
RheFlex® fittings provide inert nonmetal connections to 10-32 internal thread ports, including all Rheodyne® injectors, valves, and sample loops
Nuts and ferrules do not bind to each other during tightening and are reusable. The standard fitting can be wrench or finger tightened, and the short fitting is wrench tightened only.
The 1/16" fitting will hold PEEK, stainless steel, and titanium tubing to 5000psi.
Ordering information: Set contains five nuts and five ferrules.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty