Would you like to log in again?
Rheodyne® Inlet Filters, IDEX Health & Science
Supplier: IDEX Health & Science
7315-010 7335-010
50806-987PK 315.16 USD InStock
50806-987 50806-983
Rheodyne® Inlet Filters, IDEX Health & Science
Filters Liquid Process Filters
Inlet filters are used to extract particles from the sample between the injector and the column.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty