Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Inoculating Loops, Platinum, McCormick Scientific

Supplier: Leica
39101039 39101021 39101138 39101054
50807-020EA 325.52 USD InStock
50807-020 50807-085 50807-041 50807-008
Inoculating Loops, Platinum, McCormick Scientific
Spreaders Inoculating Loops and Needles
Made of platinum wire with 15% iridium.

  • Fused loops design
  • Platinum and iridium construction
  • Inner diameter: 2-5 mm

Loops are 45 mm in length.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty