Would you like to log in again?
Rhino VET 360 Veterinary Refractometer, Reichert
Supplier: Reichert
137536L0
51100-100EA 534.77 USD InStock
51100-100
Rhino VET 360 Veterinary Refractometer, Reichert
Refractometers Handheld Refractometers
The Rhino-rugged water resistant, dustproof, and shock resistant hand-held refractometer uses Automatic Temperature Compensation to provide accurate readings regardless of ambient temperature.
  • Automatically corrects all readings back to the Standard Reference Temperature of 68°F (20°C) providing greater accuracy
  • Improved optics that are easy to read
  • No batteries required making it reliable any time
  • Ideal for use in the office or outdoors
  • Large and small animal scales in one instrument
  • IP67 rated, sealed internal optics cannot be contaminated
  • Test takes only seconds
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty